Imipenem monohydrate, 98%
Imipenem monohydrate, 98%
Imipenem monohydrate, 98%
Imipenem monohydrate, 98%
Imipenem monohydrate, 98%
Imipenem monohydrate,Tienam, Primaxin, CAS # 74431-23-5 is a broad-spectrum carbapenem antibiotic that interferes with the activity of penicillin binding proteins along the inner cell wall.
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number FSA455322500
Price (MYR)
1,769.00
EA
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (MYR)
1,769.00
EA
Add to cart
Specifications
Chemical Name or MaterialImipenem monohydrate
TypeImipenem Monohydrate
CAS74431-23-5
Melting Point193.0°C to 198.0°C
Infrared SpectrumAuthentic
View more
General description
• Imipenem monohydrate is a beta-lactam derived from the compound thienamycin, produced by Streptomyces cattleya
• Disrupts cell wall integrity, preventing cell division and promoting lysis by interfering with the activity of penicillin binding proteins
• Imipenem is a third-generation cephalosporin that can kill both gram-negative and gram-positive bacteria
Applications
• Suitable for the study of antibiotic resistance
• High-quality, laboratory grade for use in vitro and in vivo experiments
RUO – Research Use Only
General References:
  1. Pericàs, J.M.; Moreno, A.; Almela, M.; García-de-la-Mària, C.; Marco, F.; Muñoz, P.; Peña, C. de Alarcón, A.; Del Río, A.; Eworo, A.; Cruceta, A.; Paré, J.C.; Mestres, C.A.; Miró, J.M. FOSIMI Investigators. Efficacy and safety of fosfomycin plus imipenem versus vancomycin for complicated bacteraemia and endocarditis due to methicillin-resistant Staphylococcus aureus: a randomized clinical trial. Clin Microbiol Infect. 2018, 24(6), 673-676.
  2. Chavan, V.V.; Dalal, A.; Nagaraja, S.; Thekkur, P.; Mansoor, H.; Meneguim, A.; Paryani, R.; Singh, P.; Kalon, S.; Das, M.; Ferlazzo, G.; Isaakidis, P. Ambulatory management of pre- and extensively drug resistant tuberculosis patients with imipenem delivered through port-a-cath: A mixed methods study on treatment outcomes and challenges. PLoS One. 2020, 16;15(6), e0234651.