Gentamicin sulfate
Gentamicin sulfate
Gentamicin sulfate
Gentamicin sulfate
Gentamicin sulfate, CAS # 1405-41-0, is an aminoglycoside mainly used to inhibit the growth of Gram-negative bacteria.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number FSA455310250
Price (MYR)
4,026.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
4,026.00
EA
Add to cart
Specifications
Chemical Name or MaterialGentamicin sulfate
TypeGentamicin Sulfate
CAS1405-41-0
Health Hazard 1Respiratory sensitisation (category 1), Skin sensitisation (category 1)
Health Hazard 2May cause allergy or asthma symptoms or breathing difficulties if inhaled, May cause an allergic skin reaction
View more
General Description
  • Gentamicin sulfate is a bactericidal aminoglycoside antibiotic which shows activity against Gram-negative bacteria
  • It works by inhibiting protein biosynthesis through binding the bacterial ribosome 30S subunit
Applications
  • Gentamicin sulfate has been used in studies to determine wound healing due to its antibiotic and antifungal properties
  • It has laboratory research applications in the study of a number of clinical conditions, including actinomycotic infection, appendicitis, cystic fibrosis, osteomyelitis, pelvic inflammatory disease, plague, gonorrhea, and tularemia
RUO – Research Use Only