Cloxacillin sodium salt monohydrate
Cloxacillin sodium salt monohydrate
Cloxacillin sodium salt monohydrate
Cloxacillin sodium salt monohydrate
Cloxacillin sodium salt monohydrate
Cloxacillin sodium salt monohydrate, CAS # 7081-44-9, is a compound that targets gram-positive bacteria and shows antibacterial activity.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number FSA455300050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCloxacillin sodium salt monohydrate
TypeCloxacillin Sodium Salt Monohydrate
CAS7081-44-9
Health Hazard 1Danger
Health Hazard 2GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled.
View more
General Description
• Cloxacillin sodium salt monohydrate is a chlorinated derivative of oxacillin that shows β-lactamase-resistant antibiotic activity
• This compound inhibits the last stage of bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs), which results in cell lysis, that is mediated by bacterial cell wall autolytic enzymes
Applications
• Cloxacillin sodium salt monohydrate targets gram-positive bacteria, and is effective against staphylococci that produce β-lactamase
• It acts in vitro against both penicillin gram-resistant and gram-sensitive staphylococci, certain streptococci, and pneumococci
• This compound is commonly used in vivo and in vitro studies as AmpC β-lactamase inhibitor in combination with β-lactam antibiotics
RUO – Research Use Only
General References:
  1. A New Antibacterial Agent: Sodium Cloxacillin Monohydrate (Tegopen). Journal of the American Medical Association. 1966, 195, 569-570.
  2. Harris, P.N.; Ferguson, J.K. Antibiotic therapy for inducible AmpC β-lactamase-producing Gram-negative bacilli: what are the alternatives to carbapenems, quinolones and aminoglycosides? Int J Antimicrob Agents. 2012, 40, (4), 297-305.