Cefoxitin sodium salt, 94%
Cefoxitin sodium salt, 94%
Cefoxitin sodium salt, 94%
Cefoxitin sodium salt, 94%
Cefoxitin sodium salt, 94%
Cefoxitin sodium salt, CAS # 33564-30-6, is a β-lactam, second-generation cephalosporin compound showing bactericidal activity against Bacteroides, gram-negative bacteria, including strains of P. mirabilis, P. vulgaris, E. coli, S. typhimurium, and S. enteritidis. | CAS: 33564-30-6 | C16H16N3NaO7S2 | 449.428 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA455280050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCefoxitin sodium salt
TypeCefoxitin Sodium Salt
CAS33564-30-6
Health Hazard 1Warning
Health Hazard 2GHS H Statement May cause an allergic skin reaction.
View more
General Description
  • Cefoxitin sodium salt is resistant to β-lactamase
  • It binds and inactivates penicillin-binding proteins (PBP) located on the inner membrane of the bacterial cell wall thus interfering with the cross-linkage of peptidoglycan chains necessary for bacterial cell wall strength and rigidity
Applications
  • Cefoxitin sodium salt is the sodium salt form of cefoxitin derived from cephamycin C
  • It is a β-lactam, second-generation cephalosporin compound with bactericidal activity against numerous species such as Bacteroides, gram-negative bacteria, including strains of P. mirabilis, P. vulgaris, E. coli, S. typhimurium, and S. enteritidis
  • Cefoxitin increases survival in mice infected with S. aureus, S. pyogenes, and K. pneumoniae
RUO – Research Use Only
General References:
  1. Brogden, R.; Heel, R.; Speight, T.; Avery, G. Cefoxitin: a review of its antibacterial activity, pharmacological properties and therapeutic use. Drugs. 1979, 17 (1), 1-37.
  2. Miller, A.K., Celozzi, E., Kong, Y., et al. Cefoxitin, a semisynthetic cephamycin antibiotic: In vivo evaluation. Antimicrob. Agents Chemother. 1974, 5, (1), 33-37.