Cefotaxime, 98%
Cefotaxime, 98%
Cefotaxime, 98%
Cefotaxime, 98%
Cefotaxime, CAS # 63527-52-6, is a beta-lactam antibiotic that has activity against gram-negative and positive bacteria
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number FSA455270050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCefotaxime
TypeCefotaxime
CAS63527-52-6
Health Hazard 1Danger
Health Hazard 2GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled.
View more
General Description
  • Cefotaxime, is an antibiotic with broad-spectrum activity against aerobic and anaerobic bacteria
  • It has been shown to have a high frequency of resistance mechanisms against strains such as Pseudomonas aeruginosa
Applications
  • L Cefotaxime has broad spectrum antibacterial activity
  • It binds to the penicillin-binding proteins (PBPs), resulting in inhibition of the final transpeptidation step of peptidoglycan synthesis in bacterial cell walls
RUO – Research Use Only
General References:
  1. Kern, W V., et al. Oral versus intravenous empirical antimicrobial therapy for fever in patients with granulocytopenia who are receiving cancer chemotherapy. The New England journal of medicine. 1999, 341(5), 312-8.