Penicillin V potassium salt, 94+%
Penicillin V potassium salt, 94+%
Penicillin V potassium salt, 94+%
Penicillin V potassium salt, 94+%
Penicillin V potassium salt, 94+%
CAS: 132-98-9 | C16H17KN2O5S | 388.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA455230050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPenicillin V potassium salt
TypePenicillin V Potassium Salt
CAS132-98-9
Health Hazard 1Danger
Health Hazard 2GHS H Statement Harmful if swallowed. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled.