Amoxicillin, 96%
Amoxicillin, 96%
Amoxicillin, 96%
Amoxicillin, 96%
CAS: 26787-78-0 | C16H19N3O5S | 365.404 g/mol
Have Questions?
Catalog NumberQuantity
FSA45514025025 g
Catalog number FSA455140250
Price (MYR)
1,682.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,682.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmoxicillin
TypeAmoxicillin
Physical FormSolid
CAS26787-78-0
Health Hazard 1Danger