Amoxicillin, 96%
Amoxicillin, 96%
Amoxicillin, 96%
Amoxicillin, 96%
CAS: 26787-78-0 | C16H19N3O5S | 365.404 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number FSA455140010
Price (MYR)
305.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
305.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmoxicillin
TypeAmoxicillin
CAS26787-78-0
Health Hazard 1Danger
Health Hazard 2GHS H Statement May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled.