Bismuth(III) sulfide, 99%, pure
Bismuth(III) sulfide, 99%, pure
Bismuth(III) sulfide, 99%, pure
Bismuth(III) sulfide, 99%, pure
Bismuth(III) sulfide, 99%, pure
Bismuth(III) sulfide, 99%, pure
CAS: 1345-07-9 | Bi2S3 | 514.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number FSA453991000
Price (MYR)
545.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
545.00
EA
Add to cart
Specifications
Chemical Name or MaterialBismuth(III) sulfide
CAS1345-07-9
Quantity100 g
EINECS Number215-716-
Assay Percent Range80.1 to 82.5% (Bi) (Complexometry)
View more
Suitable for battery materials development.
RUO – Research Use Only