Cobalt, 99+%, Thermo Scientific Chemicals
Cobalt, 99+%, Thermo Scientific Chemicals
Cobalt, 99+%, Thermo Scientific Chemicals
Cobalt, 99+%, Thermo Scientific Chemicals
Cobalt, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Catalog number FSA453912500
Price (MYR)
1,252.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
1,252.00
EA
Add to cart
Specifications
Chemical Name or MaterialCobalt
CAS7440-48-4
Melting Point1495.0°C
Boiling Point2870.0°C
Health Hazard 1Danger