Graphene, nanoplatelets 2-10 nm
Graphene, nanoplatelets 2-10 nm
Graphene, nanoplatelets 2-10 nm
Graphene, nanoplatelets 2-10 nm
Graphene, nanoplatelets 2-10 nm
CAS: 1034343-98-0 | Cx | 16.04 g/mol
Have Questions?
Catalog NumberQuantity
FSA45316050050 g
Catalog number FSA453160500
Price (MYR)
1,518.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
1,518.00
EA
Add to cart
Specifications
Chemical Name or MaterialGraphene
CAS1034343-98-0
Melting Point3650.0°C
Molecular FormulaCx
MDL NumberMFCD00144065