Dibutyl succinate, 99%
Dibutyl succinate, 99%
Dibutyl succinate, 99%
Dibutyl succinate, 99%
Dibutyl succinate, 99%
CAS: 141-03-7 | C12H22O4 | 230.3 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Catalog number FSA453011000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialDibutyl succinate
CAS141-03-7
Melting Point-29.0°C
Density0.9800g/mL
EINECS Number205-449-8