Methyl Arachidate, 99%
Methyl Arachidate, 99%
Methyl Arachidate, 99%
Methyl Arachidate, 99%
Methyl Arachidate, 99%
CAS: 1120-28-1 | C21H42O2 | 326.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA452820010
Price (MYR)
588.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
588.00
EA
Add to cart
Specifications
Chemical Name or MaterialMethyl arachidate
CAS1120-28-1
Melting Point45.0°C to 48.0°C
EINECS Number214-304-8
Boiling Point215.0°C to 216.0°C (10.0 mmHg)