L-Alanine, 99%, specified according to the requirements of Ph.Eur.
L-Alanine, 99%, specified according to the requirements of Ph.Eur.
L-Alanine, 99%, specified according to the requirements of Ph.Eur.
L-Alanine, 99%, specified according to the requirements of Ph.Eur.
Thermo Scientific Chemicals
L-Alanine, 99%, specified according to the requirements of Ph.Eur.
CAS: 56-41-7 | C3H7NO2 | 89.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number FSA451852500
Price (MYR)
540.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
540.00
EA
Add to cart
Specifications
Chemical Name or MaterialL-Alanine
Melting Point314.0°C to 316.0°C
Name Notespecified according to the requirements of Ph.Eur.
CAS56-41-7
Assay Percent Range99%