Tetrabutylammonium fluoride, 75% solution in water
CAS: 429-41-4 | C16H36FN | 261.47 g/mol
Have Questions?
Change view
Quantity:
100 g
25 g
Packaging:
View all
Glass Bottle
Catalog number FSA451801000
Price (MYR)
805.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
805.00
EA
Add to cart
Specifications
Chemical Name or MaterialTetrabutylammonium Fluoride
CAS429-41-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement Toxic if swallowed. Causes severe skin burns and eye damage.
Density0.9530g/mL