5-Bromo-1H-benzimidazole, 97%
5-Bromo-1H-benzimidazole, 97%
5-Bromo-1H-benzimidazole, 97%
5-Bromo-1H-benzimidazole, 97%
Thermo Scientific Chemicals
5-Bromo-1H-benzimidazole, 97%
CAS: 4887-88-1 | C7H5BrN2 | 197.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA451670050
Price (MYR)
668.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
668.00
EA
Add to cart
Specifications
Chemical Name or Material5-Bromo-1H-benzimidazole
CAS4887-88-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Health Hazard 3GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid contact during pregnancy/while nursing.