5-Methylpyrimidine, 98%, Thermo Scientific™
5-Methylpyrimidine, 98%, Thermo Scientific™
5-Methylpyrimidine, 98%, Thermo Scientific™
5-Methylpyrimidine, 98%, Thermo Scientific™
Thermo Scientific Chemicals
5-Methylpyrimidine, 98%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA450730010
Price (MYR)
1,181.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,181.00
EA
Add to cart
Specifications
Chemical Name or Material5-Methylpyrimidine
CAS2036-41-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep away fr