2,6-Dichloroindophenol, sodium salt hydrate, 90+%
2,6-Dichloroindophenol, sodium salt hydrate, 90+%
2,6-Dichloroindophenol, sodium salt hydrate, 90+%
2,6-Dichloroindophenol, sodium salt hydrate, 90+%
Thermo Scientific Chemicals
2,6-Dichloroindophenol, sodium salt hydrate, 90+%
CAS: 1266615-56-8 | C12H6Cl2NNaO2 | 290.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA450700250
Price (MYR)
861.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
861.00
EA
Add to cart
Specifications
Chemical Name or Material2, 6-Dichloroindophenol, sodium salt hydrate
CAS1266615-56-8
Assay Percent Range90+%
PackagingGlass bottle
Molecular FormulaC12H6Cl2NNaO2