Hydrochloric acid, 37%, for analysis, (max. 0.000001% Hg)
Hydrochloric acid, 37%, for analysis, (max. 0.000001% Hg)
Hydrochloric acid, 37%, for analysis, (max. 0.000001% Hg)
Hydrochloric acid, 37%, for analysis, (max. 0.000001% Hg)
Thermo Scientific Chemicals
Hydrochloric acid, 37%, for analysis, (max. 0.000001% Hg)
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA4505500252.5 LGlass Bottle
Catalog number FSA450550025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialHydrochloric Acid
CAS7647-01-0
Melting Point-35.0°C
Boiling Point57.0°C
Density1.1800g/mL