Check Order Status
1,4-Dioxane, 99.8%, Extra Dry, Stabilized
Element Search
Structure Search
Thermo Scientific Chemicals
1,4-Dioxane, 99.8%, Extra Dry, Stabilized
CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
FSA450171000
100 mL
AcroSeal™ Glass Bottle
Catalog number
FSA450171000
Price (MYR)
325.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
325.00
EA
Add to cart
Specifications
Chemical Name or Material
1,4-Dioxane
pH
6 to 8
Melting Point
12.0°C
CAS
123-91-1
Color
Colorless