Ciprofloxacin, 98%
Ciprofloxacin, 98%
Ciprofloxacin, 98%
Thermo Scientific Chemicals
Ciprofloxacin, 98%
Ciprofloxacin, CAS# 85721-33-1, is a second-generation fluoroquinolone showing antibiotic activity commonly used against susceptible organisms mainly gram-negative bacteria.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass Bottle
Catalog number FSA449620250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCiprofloxacin
CAS85721-33-1
Assay Percent Range97.5% min. (HPLC)
PackagingGlass Bottle
Molecular FormulaC17H18FN3O3
View more
General Information
• Ciprofloxacin is considered a synthetic broad-spectrum second-generation compound with antibiotic activity mainly against gram-negative bacteria
• It inhibits bacterial DNA gyrase and topoisomerase IV, enzymes that are essential for DNA replication, cell division, and causes double-strand breaks in the bacterial chromosome
Applications
• Ciprofloxacin presents more activity against gram-negative bacteria than gram-positive bacteria.
• Ciprofloxacin does not present cross-resistance between fluoroquinolones and other antibiotics classes, which is beneficial in laboratory experiments when other antibiotics are no longer effective
RUO – Research Use Only
General References:
  1. Nix, D. E.; DeVito, J. M. Ciprofloxacin and norfloxacin, two fluoroquinolone antimicrobials. Clin Pharm. 1987, 6, (2), 105-17.
  2. Sen, et al. Anxiogenic potential of ciprofloxacin and norfloxacin in rats. Singapore Med. J. 2007, 48, (11), 1028-30.