Atenolol, 98%
Atenolol, 98%
Atenolol, 98%
Thermo Scientific Chemicals
Atenolol, 98%
CAS: 29122-68-7 | C14H22N2O3 | 266.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA449550250
Price (MYR)
2,590.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,590.00
EA
Add to cart
Specifications
Chemical Name or MaterialAtenolol
CAS29122-68-7
Melting Point146.0°C to 148.0°C
EINECS Number249-451-7
Assay Percent Range98%