5-Hexenoic acid, 98%
5-Hexenoic acid, 98%
5-Hexenoic acid, 98%
5-Hexenoic acid, 98%
Thermo Scientific Chemicals
5-Hexenoic acid, 98%
CAS: 1577-22-6 | C6H10O2 | 114.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA449520010
Price (MYR)
410.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
410.00
EA
Add to cart
Specifications
Chemical Name or Material5-Hexenoic acid
CAS1577-22-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately