Methyl hexanoate, 99%
Methyl hexanoate, 99%
Methyl hexanoate, 99%
Methyl hexanoate, 99%
Thermo Scientific Chemicals
Methyl hexanoate, 99%
CAS: 106-70-7 | C7H14O2 | 130.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Catalog number FSA449242500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl hexanoate
CAS106-70-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.