Furosemide, 97+%
Furosemide, 97+%
Furosemide, 97+%
Thermo Scientific Chemicals
Furosemide, 97+%
CAS: 54-31-9 | C12H11ClN2O5S | 330.74 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA448970050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialFurosemide
CAS54-31-9
Health Hazard 1GHS Signal Word: Danger
Melting Point218.0°C to 220.0°C
EINECS Number200-203-6