Cortisone 21-acetate, 98+%, Thermo Scientific Chemicals
Cortisone 21-acetate, 98+%, Thermo Scientific Chemicals
Cortisone 21-acetate, 98+%, Thermo Scientific Chemicals
Cortisone 21-acetate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cortisone 21-acetate, 98+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA448960050
Price (MYR)
996.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
996.00
EA
Add to cart
Specifications
Chemical Name or MaterialCortisone 21-acetate
CAS50-04-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Suspected of damaging fertility or the unborn child.
Health Hazard 3GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.