Prednisone, 98%, Thermo Scientific Chemicals
Prednisone, 98%, Thermo Scientific Chemicals
Prednisone, 98%, Thermo Scientific Chemicals
Prednisone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Prednisone, 98%, Thermo Scientific Chemicals
Prednisone, 98%, CAS # 53-03-2, is a synthetic glucocorticoid with anti-inflammatory, immunomodulatory, antineoplastic, and adrenergic activities.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
View all
Glass Bottle
Catalog number FSA448950250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPrednisone
CAS53-03-2
Melting Point225.0°C to 229.0°C
Assay Percent Range98%
PackagingGlass Bottle
View more
General Description
• Prednisone is a synthetic glucocorticoid with anti-inflammatory and immunosuppressive activities
• It shows antineoplastic and adrenergic activities against hormone-sensitive cancer cells
• This compound binds to its receptors and enters the nucleus activating specific nuclear receptors, resulting in an altered gene expression and inhibiting proinflammatory cytokine production
Applications
• Prednisone can reduce circulating lymphocytes, promote cell differentiation, and stimulate apoptosis in sensitive tumor cell lines
• It is considered a prodrug that is converted by the liver into prednisolone which is the active steroid compound
• It is also used in some studies to investigate the effects of daily administration of MIDD0301, on the animal and organs weights and hematological profile
RUO – Research Use Only
General References:
  1. Zahn, N.M.; Huber, A.T.; Mikulsky, B.N.; Stepanski, M.E.; Kehoe, A.S.; Li, G.; Schussman, M.; Rashid Roni, M.S.; Kodali, R.; Cook, J.M.; Stafford, D.C.; Steeber, D.A.; Arnold, L.A. MIDD0301 - A first-in-class anti-inflammatory asthma drug targets GABAA receptors without causing systemic immune suppression. Basic Clin Pharmacol Toxicol. 2019, 125(1), 75-84.