4'-Isobutylacetophenone, 97%
4'-Isobutylacetophenone, 97%
4'-Isobutylacetophenone, 97%
4'-Isobutylacetophenone, 97%
Thermo Scientific Chemicals
4'-Isobutylacetophenone, 97%
CAS: 38861-78-8 | C12H16O | 176.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Catalog number FSA448820500
Price (MYR)
573.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
573.00
EA
Add to cart
Specifications
Chemical Name or Material4'-Isobutylacetophenone
CAS38861-78-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin