Ammonium molybdate, 99.99%, (trace metal basis)
Ammonium molybdate, 99.99%, (trace metal basis)
Ammonium molybdate, 99.99%, (trace metal basis)
Ammonium molybdate, 99.99%, (trace metal basis)
Thermo Scientific Chemicals
Ammonium molybdate, 99.99%, (trace metal basis)
CAS: 13106-76-8 | H8MoN2O4 | 196.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA448541000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium molybdate
CAS13106-76-8
Health Hazard 1GHS Signal Word: Warning
Quantity100 g
EINECS Number236-031-3