N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™, package of 4x25ML bottles
N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™, package of 4x25ML bottles
N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™, package of 4x25ML bottles
N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™, package of 4x25ML bottles
Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™, package of 4x25ML bottles
CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA448381000100 mLAcroSeal™ Glass Bottle
Catalog number FSA448381000
Price (MYR)
345.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
345.00
EA
Add to cart
Specifications
Chemical Name or MaterialN,N-Dimethylformamide
Melting Point-61.0°C
CAS68-12-2
Health Hazard 1Danger
Density0.9450g/mL