Sodium sulfite, 98%, for biochemistry
Sodium sulfite, 98%, for biochemistry
Sodium sulfite, 98%, for biochemistry
Sodium sulfite, 98%, for biochemistry
Thermo Scientific Chemicals
Sodium sulfite, 98%, for biochemistry
CAS: 7757-83-7 | Na2O3S | 126.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Catalog number FSA447952500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium sulfite
Melting Point>500.0°C
CAS7757-83-7
Quantity250 g
EINECS Number231-821-4