Benzyl alcohol, ACS reagent
Benzyl alcohol, ACS reagent
Benzyl alcohol, ACS reagent
Benzyl alcohol, ACS reagent
Benzyl alcohol, ACS reagent
CAS: 100-51-6 | C7H8O | 108.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA447721000100 mLGlass Bottle
Catalog number FSA447721000
Price (MYR)
322.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
322.00
EA
Add to cart
Specifications
Chemical Name or MaterialBenzyl alcohol
CAS100-51-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/phys