Nitrobenzene, 99.5%, for analysis
Nitrobenzene, 99.5%, for analysis
Nitrobenzene, 99.5%, for analysis
Nitrobenzene, 99.5%, for analysis
Thermo Scientific Chemicals
Nitrobenzene, 99.5%, for analysis
CAS: 98-95-3 | C6H5NO2 | 123.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA447652500
Price (MYR)
342.00
EA
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
342.00
EA
Add to cart
Specifications
Chemical Name or MaterialNitrobenzene
CAS98-95-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Toxic in contact with skin.
Harmful to aquatic life with long lasting effects.
Toxic if sw
Health Hazard 3GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove