Xylenol orange, sodium salt, ACS reagent
Xylenol orange, sodium salt, ACS reagent
Xylenol orange, sodium salt, ACS reagent
Xylenol orange, sodium salt, ACS reagent
Thermo Scientific Chemicals
Xylenol orange, sodium salt, ACS reagent
CAS: 3618-43-7 | C31H28N2Na4O13S | 760.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA447600250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialXylenol orange, sodium salt
CAS3618-43-7
Melting Point210°C
EINECS Number222-805-8
PackagingGlass bottle
View more
Indicator
RUO – Research Use Only