Gallic acid monohydrate, ACS reagent
Gallic acid monohydrate, ACS reagent
Gallic acid monohydrate, ACS reagent
Gallic acid monohydrate, ACS reagent
Thermo Scientific Chemicals
Gallic acid monohydrate, ACS reagent
CAS: 5995-86-8 | C7H5O5 | 169.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number FSA447581000
Price (MYR)
398.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
398.00
EA
Add to cart
Specifications
Chemical Name or MaterialGallic acid monohydrate
CAS5995-86-8
Melting Point251.0°C
Molecular FormulaC7H5O5
Linear Formula(HO)3C6H2CO2H·H2O