Crystal violet, ACS reagent
Crystal violet, ACS reagent
Crystal violet, ACS reagent
Crystal violet, ACS reagent
Thermo Scientific Chemicals
Crystal violet, ACS reagent
CAS: 548-62-9 | C25H30ClN3 | 407.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass Bottle
Catalog number FSA447570500
Price (MYR)
306.00
EA
Add to cart
Quantity:
50 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
306.00
EA
Add to cart
Chemical Identifiers
CAS548-62-9, 90-94-8
Specifications
Chemical Name or MaterialCrystal Violet
CAS548-62-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye damage.
Suspected of causing cancer.
CAS90-94-8
View more
Indicator
RUO – Research Use Only