2-Methoxyethanol, ACS reagent
2-Methoxyethanol, ACS reagent
2-Methoxyethanol, ACS reagent
2-Methoxyethanol, ACS reagent
Thermo Scientific Chemicals
2-Methoxyethanol, ACS reagent
CAS: 109-86-4 | C3H8O2 | 76.09 g/mol
Have Questions?
Catalog NumberQuantity
FSA4474800252.5 L
Catalog number FSA447480025
Price (MYR)
769.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
769.00
EA
Add to cart
Specifications
Chemical Name or Material2-Methoxyethanol
CAS109-86-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
May damage fertility.
May damage the unborn child.
Harmful if swallowed.
Flammable liquid and vapor.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/sh