Propionic acid, ACS reagent
Propionic acid, ACS reagent
Propionic acid, ACS reagent
Propionic acid, ACS reagent
Thermo Scientific Chemicals
Propionic acid, ACS reagent
CAS: 79-09-4 | C3H6O2 | 74.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Catalog number FSA447231000
Price (MYR)
363.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
363.00
EA
Add to cart
Specifications
Chemical Name or MaterialPropionic acid
CAS79-09-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
May be corrosive to metals.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p