Ammonium phosphate, dibasic, 99%, for biochemistry
Ammonium phosphate, dibasic, 99%, for biochemistry
Ammonium phosphate, dibasic, 99%, for biochemistry
Ammonium phosphate, dibasic, 99%, for biochemistry
Thermo Scientific Chemicals
Ammonium phosphate, dibasic, 99%, for biochemistry
CAS: 7783-28-0 | H9N2O4P | 132.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA447142500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium phosphate, dibasic
CAS7783-28-0
Health Hazard 1Warning
Quantity250 g
EINECS Number231-987-8