N,N-Dimethylformamide, 99.9%, for biochemistry, AcroSeal™
N,N-Dimethylformamide, 99.9%, for biochemistry, AcroSeal™
N,N-Dimethylformamide, 99.9%, for biochemistry, AcroSeal™
N,N-Dimethylformamide, 99.9%, for biochemistry, AcroSeal™
Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.9%, for biochemistry, AcroSeal™
CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
AcroSeal™ Glass bottle
Catalog number FSA447090010
Price (MYR)
529.00
EA
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
529.00
EA
Add to cart
Specifications
Chemical Name or MaterialN,N-Dimethylformamide
CAS68-12-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May damage the unborn child.
Flammable liquid and vapour.
Health Hazard 3GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wea