Isopropanol, 99%, for biochemistry and histology, AcroSeal™
CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change view
Quantity:
2.5 L
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA447080025
Price (MYR)
1,368.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
1,368.00
EA
Add to cart
Specifications
Chemical Name or MaterialIsopropanol
Melting Point-89.5°C
Absorbance0.01 max. at 300nm, 0.03 max. at 260nm, 0.05 max. at 240nm, 0.15 max. at 230nm, 0.3 max. at 220nm, 1 max. at 205nm
CAS67-63-0
Health Hazard 1Danger