tert-Butanol, 99.5%, for analysis
tert-Butanol, 99.5%, for analysis
tert-Butanol, 99.5%, for analysis
tert-Butanol, 99.5%, for analysis
Thermo Scientific Chemicals
tert-Butanol, 99.5%, for analysis
CAS: 75-65-0 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Plastic Bottle
Catalog number FSA447010025
Price (MYR)
721.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
721.00
EA
Add to cart
Specifications
Chemical Name or Materialtert-Butanol
Melting Point25.0°C to 25.5°C
CAS75-65-0
Health Hazard 1Danger
Density0.7800g/mL