Rhodamine WT, 20% solution in water
Rhodamine WT, 20% solution in water
Rhodamine WT, 20% solution in water
Rhodamine WT, 20% solution in water
Thermo Scientific Chemicals
Rhodamine WT, 20% solution in water
CAS: 37299-86-8 | C29H29ClN2Na2O5 | 566.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number FSA446971000
Price (MYR)
433.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
433.00
EA
Add to cart
Chemical Identifiers
CAS37299-86-8, 7732-18-5
Specifications
Chemical Name or MaterialRhodamine WT
CAS37299-86-8
CAS7732-18-5
Molecular FormulaC29H29ClN2Na2O5
Solubility InformationSolubility in water: miscible.