Ammonium dichromate, ACS reagent
Ammonium dichromate, ACS reagent
Ammonium dichromate, ACS reagent
Ammonium dichromate, ACS reagent
Thermo Scientific Chemicals
Ammonium dichromate, ACS reagent
CAS: 7789-09-5 | H8Cr2N2O7 | 252.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA446921000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium dichromate
Melting Point170.0°C
CAS7789-09-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful in contact with skin.
Very toxic to aquatic life with long lasting effects.
May damage fertility.
May damage the unborn child.
May cause genetic defects.
Causes damage to organs through prolonge