1,3,5-Triformylbenzene, 98%
1,3,5-Triformylbenzene, 98%
1,3,5-Triformylbenzene, 98%
1,3,5-Triformylbenzene, 98%
Thermo Scientific Chemicals
1,3,5-Triformylbenzene, 98%
CAS: 3163-76-6 | C9H6O3 | 162.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA446550050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material1, 3, 5-Triformylbenzene
CAS3163-76-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for