Calcium acetate monohydrate, ACS reagent
Calcium acetate monohydrate, ACS reagent
Calcium acetate monohydrate, ACS reagent
Calcium acetate monohydrate, ACS reagent
Thermo Scientific Chemicals
Calcium acetate monohydrate, ACS reagent
CAS: 5743-26-0 | C4H6CaO4·H2O | 176.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number FSA446355000
Price (MYR)
639.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
639.00
EA
Add to cart
Specifications
Chemical Name or MaterialCalcium acetate monohydrate
Melting Point160.0°C
CAS5743-26-0
Quantity500 g
Molecular FormulaC4H6CaO4·H2O