Citric acid, trisodium salt dihydrate, ACS reagent
CAS: 6132-04-3 | C6H5Na3O7·2H2O | 294.09 g/mol
Have Questions?
Change view
Quantity:
250 g
1 kg
Packaging:
View all
Plastic Bottle
Catalog number FSA446332500
Price (MYR)
244.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
244.00
EA
Add to cart
Specifications
Chemical Name or MaterialCitric Acid, Trisodium Salt Dihydrate
CAS6132-04-3
Melting Point>300.0°C
PackagingPlastic Bottle
Molecular FormulaC6H5Na3O7·2H2O