Cyclohexane, ACS reagent
Cyclohexane, ACS reagent
Cyclohexane, ACS reagent
Cyclohexane, ACS reagent
Thermo Scientific Chemicals
Cyclohexane, ACS reagent
CAS: 110-82-7 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
Packaging:
Plastic drum
Catalog number FSA446280050
Price (MYR)
920.00
EA
Add to cart
Quantity:
5 L
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
920.00
EA
Add to cart
Specifications
Chemical Name or MaterialCyclohexane
CAS110-82-7
Melting Point6.5°C
Density0.7700g/mL
Boiling Point81.0°C