N,N-Dimethylformamide dimethyl acetal, ca. 97%, AcroSeal™
N,N-Dimethylformamide dimethyl acetal, ca. 97%, AcroSeal™
N,N-Dimethylformamide dimethyl acetal, ca. 97%, AcroSeal™
N,N-Dimethylformamide dimethyl acetal, ca. 97%, AcroSeal™
N,N-Dimethylformamide dimethyl acetal, ca. 97%, AcroSeal™
Thermo Scientific Chemicals
N,N-Dimethylformamide dimethyl acetal, ca. 97%, AcroSeal™
CAS: 4637-24-5 | C5H13NO2 | 119.16 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA446151000100 mLAcroSeal™ Glass bottle
Catalog number FSA446151000
Price (MYR)
527.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
527.00
EA
Add to cart
Specifications
Chemical Name or MaterialN, N-Dimethylformamide dimethyl acetal
CAS4637-24-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye damage.
May cause an allergic skin reaction.
Harmful if inhaled.
CAS149-73-5
View more
Alkylation reagent
RUO – Research Use Only