Pyrrolidine, 99.5+%, purified by redistillation, AcroSeal™
Pyrrolidine, 99.5+%, purified by redistillation, AcroSeal™
Pyrrolidine, 99.5+%, purified by redistillation, AcroSeal™
Pyrrolidine, 99.5+%, purified by redistillation, AcroSeal™
Pyrrolidine, 99.5+%, purified by redistillation, AcroSeal™
Thermo Scientific Chemicals
Pyrrolidine, 99.5+%, purified by redistillation, AcroSeal™
CAS: 123-75-1 | C4H10ClN | 107.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number FSA446141000
Price (MYR)
244.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
244.00
EA
Add to cart
Specifications
Chemical Name or MaterialPyrrolidine
CAS123-75-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi